DWK60940A-12
Kimble® 33 Expansion Borosilicate Glass Vial With Attached White Urea Cap And Ptfe-Faced White Rubber Liner
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41122001
NACRES:
NB.43
Recommended Products
material
Borosilicate glass/PTFE vial
white cap (Urea Cap and PTFE-faced White Rubber Liner)
description
15-425
packaging
pkg of 144 ea
manufacturer/tradename
Kimble® 60940A-12
capacity
12 mL
diam. × H
19 mm × 65 mm
Looking for similar products? Visit Product Comparison Guide
Related Categories
General description
The KIMBLE® 33 Expansion Borosilicate Glass Vial is a clear, screw-thread vial designed for laboratory use, offering both thermal and chemical resistance. It features an attached white urea cap that ensures a secure closure, preventing contamination and evaporation of contents. The vial is lined with a PTFE-faced white rubber liner, enhancing its sealing capabilities and compatibility with various substances. This combination makes it an ideal choice for storing samples, reagents, and other laboratory materials efficiently.
Application
Storage and transport of samples, lyophilization, contain harsh reagents or liquids.
Features and Benefits
- Closed-top white urea caps with PTFE-faced white rubber liners
- 60940A-4 complies with EPA 600 series methods
- Packaged in corrugated trays with partitions
- Converted from Type I, 33 expansion borosilicate glass tubing.
Legal Information
KIMBLE is a registered trademark of DWK Life Sciences
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service