Skip to Content
Merck
All Photos(1)

Documents

A16807

Sigma-Aldrich

3-Acetylindole

98%

Synonym(s):

3-Indolyl methyl ketone, NSC 47180, NSC 58084

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9NO
CAS Number:
Molecular Weight:
159.18
Beilstein:
122579
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

crystals

mp

188-192 °C (lit.)

SMILES string

CC(=O)c1c[nH]c2ccccc12

InChI

1S/C10H9NO/c1-7(12)9-6-11-10-5-3-2-4-8(9)10/h2-6,11H,1H3

InChI key

VUIMBZIZZFSQEE-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Indole derivatives as antitumor agents
  • Endothelin-1 antagonists
  • Oncrasin-1 derivatives as inhibitors of the C-terminal domain of RNA polymerase II with antitumor activities
  • Inhibitors of hepatitis C NS3/4A serine protease
  • Antibacterial agens against MDR Staphylococcus aureus strains
  • Antimalarial agents
  • Anti-bovine viral diarrhea virus (BVDV) agents
  • HIV-1 integrase inhibitors
  • Inhibitors of NF-κB transcription regulation related to TNF-α cytokine release

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Zong-ying Liu et al.
Bioorganic & medicinal chemistry letters, 19(15), 4167-4170 (2009-06-16)
A novel series of 1-(benzo[b]thiophen-2-yl)ethanone analogues were prepared and evaluated for enhancing BMP-2 expression. Compounds 1-5, 7, 8, 12, 13 and 16, with upregulation rate values of 35.6%, 27.9%, 39.8%, 32.0%, 37.1%, 30.2%, 28.0%, 33.5%, 22.8% and 27.3% in vitro

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service