Skip to Content
Merck
All Photos(1)

Key Documents

926205

Sigma-Aldrich

Ru(OAc)2[(S)-dtbm-SEGPHOS®]

Takasago

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C78H106O12P2Ru
CAS Number:
Molecular Weight:
1398.69
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

Quality Level

form

powder

Application

Ru(OAc)2[(S)-dtbm-SEGPHOS®] is a ruthenium catalyst for asymmetric hydrogenation of olefins.

Product can be used with our benchtop hydrogen generator, H-Genie Lite (Z744083)

Legal Information

SEGPHOS is a registered trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Selective and Scalable Synthesis of Sugar Alcohols by Homogeneous Asymmetric Hydrogenation of Unprotected Ketoses
Tindall J D, et al.
Angewandte Chemie (International Edition in English), 60(2) (2020)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service