Skip to Content
Merck
All Photos(1)

Key Documents

920991

Sigma-Aldrich

Silver(I) carbonate

≥99.9% trace metals basis

Synonym(s):

Silver carbonate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Ag2CO3
CAS Number:
Molecular Weight:
275.75
Beilstein:
6936654
MDL number:
UNSPSC Code:
12352302
NACRES:
NA.23
Assay:
≥99.9% trace metals basis
form:
powder

Quality Level

Assay

≥99.9% trace metals basis

form

powder

mp

210 °C (dec.) (lit.)

density

6.08 g/mL at 25 °C (lit.)

SMILES string

[Ag]OC(=O)O[Ag]

InChI

1S/CH2O3.2Ag/c2-1(3)4;;/h(H2,2,3,4);;/q;2*+1/p-2

InChI key

KQTXIZHBFFWWFW-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Application

Silver carbonate can be used as a catalyst to synthesize α-trifluoromethylacrylic acids and 2-(pyridin-3-yl)pyrimidines (PPMDs), key building blocks for synthesizing bioactive molecules. Additionally, Ag2CO3 can also be used to functionalize metallofullerenes with potential photovoltaic and biomedical applications.

Pictograms

CorrosionEnvironment

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1 - Eye Dam. 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Silver-catalyzed cascade reactions of 3-cyanochromone with 1, 1-enediamines: synthesis of highly functionalized 2-(pyridin-3-yl)-chromeno [2, 3-d] pyrimidines.
Xiao Q, et al.
Organic Chemistry Frontiers : An International Journal of Organic Chemistry / Royal Society of Chemistry, 7(15), 2035-2039 (2020)
Ligand Free Palladium-Catalyzed Synthesis of a-Trifluoromethylacrylic Acids and Related Acrylates by Three-Component Reaction
Xiao P, et al.
Advanced Synthesis & Catalysis, 362(4), 949-954 (2020)
Jia Li et al.
Frontiers in chemistry, 8, 593602-593602 (2020-11-17)
A novel radical reaction of monometallofullerene Y@C2v(9)-C82 with N-arylbezamidine (1) is successfully conducted through catalysis with silver carbonate. The high-performance liquid chromatographic and mass spectrum results demonstrate that the reaction is highly regioselective to afford only one monoadduct (2) with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service