Skip to Content
Merck
All Photos(1)

Documents

731803

Sigma-Aldrich

Poly(ethylene glycol) diglycidyl ether

average MN 6,000, cross-linking reagent amine reactive

Synonym(s):

Polyethylene glycol, Diepoxy PEG, PEG diglycidyl ether, Polyoxyethylene bis(glycidyl ether)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5O2-(C2H4O)n-C3H5O
CAS Number:
UNSPSC Code:
12162002
NACRES:
NA.23

product name

Poly(ethylene glycol) diglycidyl ether, Mn 6,000

form

liquid

Quality Level

mol wt

Mn 6,000

reaction suitability

reagent type: cross-linking reagent
reactivity: amine reactive

mp

57-62 °C

polymer architecture

shape: linear
functionality: homobifunctional

storage temp.

2-8°C

InChI

1S/C8H14O4/c1(9-3-7-5-11-7)2-10-4-8-6-12-8/h7-8H,1-6H2

InChI key

AOBIOSPNXBMOAT-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

  • Water-Insoluble, Thermostable, Crosslinked Gelatin Matrix for Soft Tissue Implant Development.: This study explores the use of Poly(ethylene glycol) diglycidyl ether (PEGDE) in creating thermostable, water-insoluble gelatin matrices for soft tissue implants. The research highlights PEGDE′s effectiveness as a crosslinking agent, enhancing the biocompatibility and mechanical properties of the matrix (Varga et al., 2024).
  • Alumina Incorporation in Self-Supported Poly(ethylenimine) Sorbents for Direct Air Capture.: Although not directly focused on PEGDE, this study includes it as a part of the sorbent material formulation, demonstrating its versatility in industrial applications beyond biomedical fields (Narayanan et al., 2024).
  • Lignin-Based Hydrogels for the Delivery of Bioactive Chaga Mushroom Extract.: This research utilizes PEGDE to enhance the properties of lignin-based hydrogels, which are used for controlled delivery of bioactive compounds. The study showcases PEGDE′s role in improving the hydrogel′s stability and release characteristics (Nagardeolekar et al., 2024).
  • Ductile, High-Lignin-Content Thermoset Films and Coatings.: This study investigates the use of PEGDE in developing high-lignin-content thermoset films and coatings, emphasizing its application in creating durable and flexible materials for industrial use (Boarino et al., 2023).
  • Developing Mixed Matrix Membranes with Good CO2 Separation Performance Based on PEG-Modified UiO-66 MOF and 6FDA-Durene Polyimide.: This article discusses the use of PEGDE in modifying metal-organic frameworks (MOFs) for mixed matrix membranes, highlighting its potential in enhancing CO2 separation performance, crucial for environmental and industrial applications (Veetil et al., 2023).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

386.6 °F - closed cup

Flash Point(C)

197.00 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service