Skip to Content
Merck
All Photos(1)

Key Documents

730254

Sigma-Aldrich

2,2,3,3,4,4,4-Heptafluorobutylamine

96%

Synonym(s):

1H,1H-Heptafluorobutylamine, 2,2,3,3,4,4,4-Heptafluoro-1-butanamine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C4H4F7N
CAS Number:
Molecular Weight:
199.07
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Quality Level

Assay

96%

form

liquid

density

1.456 g/mL at 25 °C

functional group

amine
fluoro

SMILES string

NCC(F)(F)C(F)(F)C(F)(F)F

InChI

1S/C4H4F7N/c5-2(6,1-12)3(7,8)4(9,10)11/h1,12H2

InChI key

WBGBQSRNXPVFDB-UHFFFAOYSA-N

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

210.0 °F

Flash Point(C)

98.9 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Esther van Andel et al.
ACS applied materials & interfaces, 9(44), 38211-38221 (2017-10-25)
Micron- and nano-sized particles are extensively used in various biomedical applications. However, their performance is often drastically hampered by the nonspecific adsorption of biomolecules, a process called biofouling, which can cause false-positive and false-negative outcomes in diagnostic tests. Although antifouling

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service