Skip to Content
Merck
All Photos(1)

Documents

728411

Sigma-Aldrich

2-tert-Butylimino-2-diethylamino-1,3-dimethylperhydro-1,3,2-diazaphosphorine solution

1 M in hexane, ≥97% (GC), 97% (GC)

Synonym(s):

BEMP

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H31N4P
CAS Number:
Molecular Weight:
274.39
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97% (GC)
97% (GC)

form

liquid

concentration

1 M in hexane

impurities

≤0.10% water

SMILES string

CCN(CC)P1(=NC(C)(C)C)N(C)CCCN1C

InChI

1S/C13H31N4P/c1-8-17(9-2)18(14-13(3,4)5)15(6)11-10-12-16(18)7/h8-12H2,1-7H3

InChI key

VSCBATMPTLKTOV-UHFFFAOYSA-N

Signal Word

Danger

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Skin Corr. 1B - STOT RE 1 Inhalation - STOT SE 3

Target Organs

Central nervous system, Nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-7.6 °F - closed cup

Flash Point(C)

-22 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service