Skip to Content
Merck
All Photos(1)

Key Documents

714372

Sigma-Aldrich

Methyl 2-chloro-4-(trifluoromethyl)pyrimidine-5-carboxylate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4ClF3N2O2
CAS Number:
Molecular Weight:
240.57
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form:
liquid
Assay:
97%
Pricing and availability is not currently available. Contact Technical Service

Quality Level

Assay

97%

form

liquid

refractive index

n20/D 1.468

density

1.491 g/mL at 25 °C

functional group

chloro
ester
fluoro

storage temp.

2-8°C

SMILES string

COC(=O)c1cnc(Cl)nc1C(F)(F)F

InChI

1S/C7H4ClF3N2O2/c1-15-5(14)3-2-12-6(8)13-4(3)7(9,10)11/h2H,1H3

InChI key

VLUBJYUOPNGBOQ-UHFFFAOYSA-N

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service