Skip to Content
Merck
All Photos(1)

Documents

687901

Sigma-Aldrich

4-(Trifluoromethoxy)phenylmagnesium bromide solution

0.5 M in THF

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H4BrF3MgO
Molecular Weight:
265.31
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

Quality Level

reaction suitability

reaction type: Grignard Reaction

concentration

0.5 M in THF

density

0.975 g/mL at 25 °C

SMILES string

FC(F)(F)Oc1ccc([Mg]Br)cc1

InChI

1S/C7H4F3O.BrH.Mg/c8-7(9,10)11-6-4-2-1-3-5-6;;/h2-5H;1H;/q;;+1/p-1

InChI key

UFMHNMPAQCQXMW-UHFFFAOYSA-M

Application

Reactant for:
  • Preparation of chromanol derivatives as novel class of CETP inhibitors for treatment of cardiovascular disease
  • Pd-catalyzed cross coupling
  • Preparation of indazole-pyridine derivatives as protein kinase B/Akt inhibitors with reduced hypotension, via Stille coupling, Mitsunobu reaction and copper-catalyzed aziridine ring-opening reaction

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-22.0 - -4.0 °F - closed cup

Flash Point(C)

-30 - -20 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service