Skip to Content
Merck
All Photos(3)

Key Documents

533068

Sigma-Aldrich

6-Methoxy-3-pyridinecarboxaldehyde

98%

Synonym(s):

6-Methoxy-3-nicotinaldehyde

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H7NO2
CAS Number:
Molecular Weight:
137.14
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
EC Index Number:
627-354-4
NACRES:
NA.22

Quality Level

Assay

98%

mp

51-54 °C (lit.)

SMILES string

COc1ccc(C=O)cn1

InChI

1S/C7H7NO2/c1-10-7-3-2-6(5-9)4-8-7/h2-5H,1H3

InChI key

CTAIEPPAOULMFY-UHFFFAOYSA-N

General description

6-Methoxy-3-pyridinecarboxaldehyde, also known as 6-methoxy-3-nicotinaldehyde, can be prepared from 5-bromo-2-methoxypyridine.

Application

6-Methoxy-3-pyridinecarboxaldehyde may be used to prepare 6-methoxy-4-methyl-3-pyridinecarboxaldehyde and 6-methoxy-5-methyl-3-pyridinecarboxaldehyde.
An intermediate for the synthesis of the somatostatic sst3 receptor.
Substrate used in a synthesis of flavanones via an L-proline-catalyzed condensation with o-hydroxyarylketones.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Tetrahedron Asymmetry, 14, 3469-3477 (2003)
Tetrahedron Letters, 46, 6991-6991 (2005)
Lithiation of methoxypyridines directed by ?-amino alkoxides.
Comins DL and Killpack MO.
The Journal of Organic Chemistry, 55(1), 69-73 (1990)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service