Skip to Content
Merck
All Photos(2)

Key Documents

335193

Sigma-Aldrich

Copper(II) hexafluoroacetylacetonate hydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Cu(C5HF6O2)2 · xH2O
CAS Number:
Molecular Weight:
477.65 (anhydrous basis)
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

powder or chunks
solid

composition

Cu, 12.0-14.0% EDTA titration
Degree of hydration, ≤1

reaction suitability

core: copper

mp

130-134 °C (lit.)

SMILES string

O.FC(F)(F)C(=O)\C=C(/O[Cu]O\C(=C/C(=O)C(F)(F)F)C(F)(F)F)C(F)(F)F

InChI

1S/2C5H2F6O2.Cu.H2O/c2*6-4(7,8)2(12)1-3(13)5(9,10)11;;/h2*1,12H;;1H2/q;;+2;/p-2/b2*2-1-;;

InChI key

SREVICRJIZZXHD-SUXDNRKISA-L

Related Categories

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ville Saarimaa et al.
ACS applied materials & interfaces, 10(25), 21730-21739 (2018-06-06)
Zinc carbonate and a mixed-phase zinc carbonate were precipitated selectively on hot dip galvanized steel in the presence of CO2 and water. The zinc carbonate was precipitated as a uniform layer with cubic superficial appearance, while the mixed-phase zinc carbonate

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service