Skip to Content

302244

Indoline-2-carboxylic acid

97%

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C9H9NO2
CAS Number:
Molecular Weight:
163.17
NACRES:
NA.22
PubChem Substance ID:
UNSPSC Code:
12352106
EC Number:
278-899-6
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

97%

reaction suitability

reaction type: solution phase peptide synthesis

mp

168 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

OC(=O)C1Cc2ccccc2N1

InChI

1S/C9H9NO2/c11-9(12)8-5-6-3-1-2-4-7(6)10-8/h1-4,8,10H,5H2,(H,11,12)

InChI key

QNRXNRGSOJZINA-UHFFFAOYSA-N



pictograms

Health hazardExclamation mark

signalword

Warning

hcodes

Hazard Classifications

Repr. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

Eyeshields, Faceshields, Gloves, type P3 (EN 143) respirator cartridges



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library