Skip to Content
Merck
All Photos(1)

Documents

253693

Sigma-Aldrich

2-Fluorophenyl isothiocyanate

98%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
FC6H4NCS
CAS Number:
Molecular Weight:
153.18
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.6254 (lit.)

density

1.248 g/mL at 25 °C (lit.)

SMILES string

Fc1ccccc1N=C=S

InChI

1S/C7H4FNS/c8-6-3-1-2-4-7(6)9-5-10/h1-4H

InChI key

OAGDRIUTLPDSMJ-UHFFFAOYSA-N

Application

2-Fluorophenyl isothiocyanate has been used in the synthesis of acylthiosemicarbazides.

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

206.6 °F - closed cup

Flash Point(C)

97 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

New Heterocyclic Compounds from 1, 2, 4-triazole and 1, 3, 4-thiadiazole Class Having Diphenylsulfone and 2-fluorophenyl Fragments.
Barbuceanu S-F, et al.
Revista de Chimie (Bucharest, Romania), 62, 308-312 (2011)
Meenakshi Verma et al.
Journal of materials chemistry. B, 8(36), 8405-8413 (2020-09-24)
Okadaic acid (OA) is one of the known marine biotoxins produced by various dinoflagellates and exists in seafood such as shellfish. The consumption of contaminated shellfish with OA leads to diarrheic shellfish poisoning (DSP), which results in the inhibition of
Fazila Rizvi et al.
Scientific reports, 9(1), 6738-6738 (2019-05-03)
A library of thiosemicarbazide derivatives of isoniazid 3-27, was synthesized and evaluated for their anti-inflammatory and urease inhibition activities, by using in vitro bioassays. Among these compounds 9, 10, 12, 21, and 26 were identified as new derivatives. Prolonged use

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service