Skip to Content
Merck
All Photos(2)

Documents

233579

Sigma-Aldrich

4-Octadecylaniline

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)17C6H4NH2
CAS Number:
Molecular Weight:
345.60
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

97%

bp

240-245 °C/0.4 mmHg (lit.)

mp

59-63 °C (lit.)

SMILES string

CCCCCCCCCCCCCCCCCCc1ccc(N)cc1

InChI

1S/C24H43N/c1-2-3-4-5-6-7-8-9-10-11-12-13-14-15-16-17-18-23-19-21-24(25)22-20-23/h19-22H,2-18,25H2,1H3

InChI key

CCRTYBGROAGVGK-UHFFFAOYSA-N

Application

4-Octadecylaniline was used in the fabrication of alternate-layer Langmuir-Blodgett (LB) films and pyroelectric activity in the films was assesd.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

IR studies of pyroelectric Langmuir-Blodgett films.
Jones CA, et al.
Thin Solid Films, 155(2), 187-195 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service