Merck
Evaluation of Xenostrip-Tv, a rapid diagnostic test for Trichomonas vaginalis infection.
Journal of clinical microbiology (2004-08-07)
A Pillay, J Lewis, R C Ballard
RÉSUMÉ
An immunochromatographic strip test, Xenostrip-Tv, was compared to wet mount and PCR for the diagnosis of Trichomonas vaginalis infection in women. Of 428 specimens tested, 54 (12.6%) were positive by an "expanded gold standard," defined as either a positive wet mount and PCR test with primers TVK3 and TVK7 and/or a positive PCR test confirmed by a second PCR assay with primers TVA5-1 and TVA6; 26 (6%) were positive by wet mount, and 36 (8.4%) were positive by Xenostrip-Tv test. Since the Xenostrip-Tv test is rapid and easy to perform and proved to be more sensitive than wet mount, it should be considered as an alternative to wet mount for point-of-care diagnosis of trichomoniasis, especially in settings where microscopy is impractical.
MATÉRIAUX
Référence du produit
Marque
Description du produit
Roche
dTTP, Lithium salt