Key Documents
58458
Supelco
Raccords Waters®
size 1/16 in., pkg of 5 ea
About This Item
Code UNSPSC :
41115700
Description
nuts and ferrules (5 ea)
Conditionnement
pkg of 5 ea
Taille
1/16 in.
Description générale
The Waters® Fittings are used for connecting most HPLC columns to a 1/16 in. tubing system.
Produits recommandés
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Informations légales
Waters is a registered trademark of Waters