Principaux documents
509396
Seringue SGE étanche aux gaz, aiguille amovible
2.5MDR-GT, volume 2500 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
Synonyme(s) :
SERINGUE SGE 2,5 ML, ÉTANCHE AUX GAZ
About This Item
Code UNSPSC :
41122001
Matériaux
bevel tip stainless steel needle (point style 2)
Taille de l'aiguille
23 ga
Caractéristiques
removable needle
Conditionnement
pkg of 1 ea
Fabricant/nom de marque
SGE 008500
Aiguille, L × D.E.
50 mm × 0.63 mm
Volume
2500 μL
Description générale
SGE gas tight syringe with removable needle, Model: 2.5MDR-GT (volume 2.5 mL, needle size 23 ga (bevel tip), needle L × O.D. 50 mm × 0.63 mm)
  • Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
  • Autoclavable syringes but the plunger should be removed.
  • Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
  • Maximum inertness for liquid or gas samples.
  • Suitable for manual dispenser/diluter applications.
  • High accuracy of dispensed volume.