Key Documents
Z711020
Media bottles, wide mouth, Lab 45, accessory
red ETFE pour ring
Synonyme(s) :
240760
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Matériaux
red ETFE pour ring
Fabricant/nom de marque
WHEATON®
Caractéristiques et avantages
• For high temperature applications, use the PBT (polybutylene terephthalate) screw caps and the ETFE (ethylene tetrafluoroethylene) pour lips which can withstand temperatures up to 200ºC up to 2 hours• Autoclavable
Informations légales
WHEATON is a registered trademark of Wheaton Industries, Inc.