Principaux documents
Z374962
PCR microtube and cap strips
capacity 0.2 mL
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
cap (strips)
colorless tube
Stérilité
sterile
Caractéristiques
neck thread push fit
Conditionnement
pkg of 250 strips (8 tubes or caps per strip)
Technique(s)
PCR: suitable
Capacité
0.2 mL
Volume de travail
0.2 mL
Type de liaison
non-treated surface
Description générale
Strips of eight tubes connected with double bridges to avoid accidental separation. Caps also are in strips of eight. Can be cut apart to use individually if desired.
Protocoles
Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.
Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.
Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.
Hot Start dNTPs block DNA polymerase until heat activation, enhancing PCR specificity.
Afficher tout