Key Documents
VM03
Sigma-Aldrich
Filter Plate
Removes cell debris prior to using the GenElute 96 Well Total RNA Kit
About This Item
Code UNSPSC :
41104923
Nomenclature NACRES :
NB.24
Niveau de qualité
Application
The Filter Plate can be used with Sigma′s GenElute 96 Well Total RNA Purification Kit to remove cell debris and prevent the RNA Binding Plate from clogging. The Filter Plate is used to filter the lysate before applying the lysate to the RNA Binding Plate (GenElute 96 Well Total RNA Purification Kit).