Principaux documents
TMO362002-0002
Nalgene® Boston round bottle without cap
natural high-density polyethylene, volume 60 mL, case of 1000 ea
Synonyme(s) :
HDPE bottle, nalgene bottle
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Matériaux
natural high-density polyethylene
Conditionnement
case of 1000 ea
Fabricant/nom de marque
Nalgene 362002-0002
Capacité
60 mL
Volume
60 mL
Description générale
Conduct demanding packaging applications with Thermo Scientific Nalgene Boston Round HDPE Bottles without Closure, which are both durable and reliable products. These high density polyethylene bottles are rigid and translucent with excellent chemical resistance. Uniform walls make these bottles resistant to splitting and puncturing.
Informations légales
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries