Accéder au contenu
MTCC3175AST
MTC™ Bio ClearSeal™ Microtubes screw caps
for use with 2.0 mL microtubes MTCC3172, assorted colors, pack of 1000 ea
Synonyme(s) :
conical tube, micro centrifuge tube, screw-cap, MTC Bio C3175-AST, sample collection tube, sample receptacle, sample tube
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.11
UNSPSC Code:
41122500
Material:
polypropylene , assorted colors
Sterility:
non-sterile
Packaging:
pack of 1000 ea
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
polypropylene , assorted colors
description
self standing
sterility
non-sterile
packaging
pack of 1000 ea
manufacturer/tradename
MTC Bio C3175-AST
General description
Screw cap for 2.0mL microtube MTC3172-1000EA, assorted colors (red, blue, green, yellow, orange), non-sterile, with installed O-ring. Packaged as 5 Individually coloured bags of 200 caps, total 1000 caps.
Application
  • General laboratory research
  • Diagnostic / Reagent packaging
Features and Benefits
  • Leak-proof O-ring seal from -86°C to +121°C
Legal Information
ClearSeal is a trademark of MTC Bio
MTC is a trademark of MTC Bio