Principaux documents
GBL702525
Grace Bio-Labs HybriSlip hybridization cover
L × W × thickness 25 mm × 25 mm × 0.25 mm, pkg of 1000 ea
Synonyme(s) :
coverslide, hydrophobic coverslip, nuclease free coverslip, thin plastic coverslip
About This Item
Code UNSPSC :
41122600
Nomenclature NACRES :
NB.22
Matériaux
colorless
polycarbonate
Conditionnement
pkg of 1000 ea
Fabricant/nom de marque
Grace Bio-Labs 702525
L × l × épaisseur
25 mm × 25 mm × 0.25 mm
Description générale
HybriSlip ideally suited for in situ hybridization, in situ PCR and hybridization to genomic arrays on glass slides.
HybriSlips are flexible, thin (0.25 mm) covers that minimize fluid friction and facilitate uniform reagent distribution. RNase free, hydrophobic covers will not trap or bind probes to their surfaces like coverglass. Product is ready-to-use without pretreatment. Working surfaces are protected with clean release liners to prevent RNase contamination. HybriSlips remain flat and will not curl, even at high temperatures.
  • RNase and DNase free
  • Hydrophobic
  • Disposable
  • Will not chip or break
Informations légales
HybriSlip is a trademark of Grace Bio-Labs, Inc.