Principaux documents
EP5910763000
Adapter for Eppendorf® Rotor S-4xUniversal-Large
0.5-2mL micro test tubes, rotor S-4xUniversal
Synonyme(s) :
Adapter for Eppendorf® S-4x Universal Large Rotor
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.15
Caractéristiques
0.5-2mL micro test tubes
Conditionnement
pack of 2 ea
Fabricant/nom de marque
5910763000
Compatibilité
rotor S-4xUniversal
Description générale
2 microtest tube adapters to enable use of 1.5 to 2 mL tubes in rotor S-4xUniversal
Adapters for various sizes of tubes and bottles for use with Eppendorf S-4x Universal large rotor.
Informations légales
Eppendorf is a registered trademark of Eppendorf AG