Principaux documents
EP5427717005
Eppendorf® Adapter
Holds PCR Strips tubes, pack of 4 ea
About This Item
Code UNSPSC :
41103911
Nomenclature NACRES :
NB.82
Caractéristiques
Holds PCR Strips tubes
Conditionnement
pack of 4 ea
Fabricant/nom de marque
Eppendorf® 5427717005
Technique(s)
PCR: suitable
centrifugation: suitable
Compatibilité
for use with Rotor F-45-64-5-PCR
Informations légales
Eppendorf is a registered trademark of Eppendorf AG