EP4421601009
Eppendorf® Membrane Filter
pore size 0.45 μm, sterile, PTFE, pkg of 5 pieces, (Easypet 4421)
Synonyme(s) :
Eppendorf Filter
About This Item
Code UNSPSC :
40161507
Nomenclature NACRES :
NB.24
Produits recommandés
Informations légales
Eppendorf is a registered trademark of Eppendorf AG