CLS6796620D
Corning® combo hotplate and stirrer with digital display
AC/DC input 230 V AC (Supplied with both 3-pin UK plug and 2-pin Euro plug), plate L × W 10 in. × 10 in., CE compliant
Synonyme(s) :
stirrers
About This Item
Produits recommandés
Caractéristiques
CE compliant
Conditionnement
case of 1
Fabricant/nom de marque
Corning 6796-620D
Disponibilité
not available in USA and Canada
Paramètres
5-550 °C temp. range
60-1150 rpm speed
Plaque/Plateau, L × l
10 in. × 10 in.
Description générale
- Digital LED temperature display is adjustable in 5 °C increments and blinks until set temperature is reached
- Microprocessor maintains consistent and repeatable temperature and stir speed settings.
- Bright LED HOT TOP icon lights up when top plate temperature is over 60 °C, even when heat control is turned off.
- Separate temperature sensor provides power cut off if unit overheats
- Extremely durable and heat resistant Pyroceram glass-ceramic top
- Small footprint and low profile
- Meets UL and cUL standards