Principaux documents
CLS6627
Corning® Step-R Repeating Pipettor Syringe Tips
volume (50.0 mL), non-sterile
Synonyme(s) :
pipet tips
About This Item
Code UNSPSC :
41121600
Nomenclature NACRES :
NB.02
Stérilité
non-sterile
Conditionnement
pack of 25 ea
Fabricant/nom de marque
Corning 6627
Volume
(50.0 mL)
Description générale
  • Ideal for viscous, dense and volatile liquides
  • Minimum increments printed on the syringe itself
  • Accurately deliver selected dose
  • Made from high quality polypropylene and polyethylene
Informations légales
Corning is a registered trademark of Corning, Inc.