Merck
CLS3294
Corning® CellBIND® surface culture dishes
D × H 35 mm × 10 mm
Synonyme(s) :
plastic cell culture dish
Se connecter pour consulter les tarifs organisationnels et contractuels.
Changer de vue
A propos de cet article
NACRES:
NB.13
UNSPSC Code:
41121812
Material:
polystyrene , round
Sterility:
sterile; γ-irradiated
Manufacturer/tradename:
Corning 3294
Packaging:
sleeve of 10
Service technique
Besoin d'aide ? Notre équipe de scientifiques expérimentés est là pour vous.
Laissez-nous vous aider
material
polystyrene , round
sterility
sterile; γ-irradiated
packaging
sleeve of 10
manufacturer/tradename
Corning 3294
D × H
35 mm × 10 mm
surface area
8 cm2
General description
  • Manufactured from optically-clear virgin polystyrene
  • Corning® CellBIND® surface treated for optimal cell attachment
  • Sterilized by gamma radiation and certified nonpyrogenic
  • Have stacking beads to aid in handling
  • Supplied with vents to provide consistent gas exchange
Legal Information
CellBIND is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.