Principaux documents
BAB617003500
H-B® Durac® Timer
Timing capacity : 99 Minute:99 Second, 10 button input
Synonyme(s) :
laboratory timer, pocket timer, timer alarm
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.79
Caractéristiques
10 button input
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Bel-Art B61700-3500
Écran, l × H
53.3 mm × 22.9 mm
Description générale
Ten button timer allows direct input for faster programming of desired time
Caractéristiques et avantages
Timing capacity: 99 minutes: 99 seconds
Count direction: up/down
Last time set memory
Audible and visual alarms; displays elapsed time after countdown
Mounting options: lay flat on bench, flip-open stand for vertical view, pocket clip for attachment to lab coat, magnetic back for metal surface attachment
Requires two AAA batteries (included)
Informations légales
Durac is a registered trademark of SP Scienceware, a division of SP
H-B is a registered trademark of SP Scienceware, a division of SP