Principaux documents
AXYPCR96C
Corning® Axygen® PCR Microplate
size 96 wells, conical bottom clear polypropylene wells, pkg of 5x10plates/cs
Synonyme(s) :
96 multiwell plate
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.22
Matériaux
conical bottom clear polypropylene wells
Stérilité
non-sterile
Caractéristiques
lid: no
skirt: no
Fabricant/nom de marque
Axygen Inc PCR-96-C
Corning
Conditionnement
pkg of 5x10plates/cs
Volume maximum
200 μL
Taille
96 wells
Volume de travail
200 μL
Adéquation
suitable for (Statagene MX4000)
Type de liaison
non-treated surface
Informations légales
Axygen is a registered trademark of Corning, Inc.
Corning is a registered trademark of Corning, Inc.