Principaux documents
Z759317
Sigma-Aldrich
Joint clip, standard taper
joint: 24, green polyethylene
Synonyme(s) :
NS joint clamps, NS joint clips, National standard joint clamps, National standard joint clips
About This Item
Code UNSPSC :
41122413
Nomenclature NACRES :
NB.43
Matériaux
green polyethylene
Joint
joint: 24
Conditionnement
pkg of 50 ea
Description générale
Color coded, rigid polyethylene joint clips fit standard taper joints. Will not scratch glassware or corrode; withstands temperatures up to 120 °C.
Les clients ont également consulté
Slide 1 of 2
1 of 2