Principaux documents
Z743249
Julabo® bath spring clamps for Erlenmeyer flasks
fits 10 mL flask
About This Item
Code UNSPSC :
41103706
Nomenclature NACRES :
NB.77
Conditionnement
pack of 1 ea
Fabricant/nom de marque
Julabo 8970601
Description générale
The individual spring clips can be positioned in the shaking water bath using the base tray for spring clips (Z743201-1EA). A maximum of 56 spring clips for 10 ml Erlenmeyer flasks can be mounted on a base tray.
Informations légales
Julabo is a registered trademark of Julabo USA, Inc.