Principaux documents
Z742442
Incu-Shaker Mini dedicated platform
for 5 x 500 mL Erlenmeyer flasks
About This Item
Code UNSPSC :
41103811
Nomenclature NACRES :
NB.73
Caractéristiques
for 5 x 500 mL Erlenmeyer flasks
Conditionnement
pkg of 1
Fabricant/nom de marque
Benchmark Scientific H1000-P-500
Description générale
Dedicated platform for the Incu-Shaker Mini that features a non-slip rubber coated surface, ideal for tissue culture flasks, petri dishes and staining trays.
Informations légales
Incu-Shaker is a trademark of Benchmark Scientific, Inc.