Principaux documents
Z740686
Sigma-Aldrich
Aldrich® Essentials filter flask, with side arm
capacity 1000 mL
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass flask
flat bottom flask
Joint
joint stopper10
Conditionnement
pkg of 6 ea
Capacité
1000 mL
Description générale
Clear borosilicate glass Erlenmeyer style flask with side arm for connecting to vacuum or aspirator line. Used for filtration with Buchner funnels, Gooch crucibles, etc. Beaded rim for rubber gromets or filter adapters or rubber stoppers.
Informations légales
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC