Z682411
Ace vapor duct tube, for condenser "A" assembly
joint: ST/NS 24/40, L 160 mm, fits Büchi® models R-210/R-215
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Produits recommandés
Joint
joint: ST/NS24/40
Fabricant/nom de marque
Büchi equivalent, 48068
Longueur
160 mm
Description générale
The vapor duct tube accessories are for Büchi® condenser "A" assemblies, and will fit either Büchi evaporator models R-210/R-215 or R-114/R-144.
Combi-clips: not included.
Combi-clips: not included.
Informations légales
Büchi is a registered trademark of Büchi Labortechnik AG