Z680869
Duran® stirred reactor caps and accessories
folding magnetic stirrer with shaft
Synonyme(s) :
1200391
About This Item
Produits recommandés
Conditionnement
pack of 1 ea
Fabricant/nom de marque
DURAN® 1200391
Raccords
for for GL 45 stirred reactor systems
Description générale
Caractéristiques et avantages
- Manufactured from chemically resistant materials: PP, PTFE, PEEK, and stainless steel
- Use with standard magnetic stirrers up to 500 rpm and allows a significantly improved mixing performance compared to magnetic stir bars
- The adjustable stirrer shaft can be moved to the optimal position in either 500 or 1000 ml DURAN® GL 45 bottles
- Flexible: Use with the DURAN® GL 45 / GL 14 tubing connection components to connect 1.6, 3.0, 3.2, or 6.0 mm outer diameter semi-rigid tubing for the addition or removal of liquids or gas
- Autoclavable
- Not suitable for use with overhead lab stirrers