Principaux documents
Z674575
GyroMini nutating mixer
AC/DC input 230 V AC, UK plug
Synonyme(s) :
lab equipment accessory
About This Item
Code UNSPSC :
41103801
Nomenclature NACRES :
NB.73
Description générale
  • Small foot print means less bench space needed
  • Gyratory action provides gentle, thorough mixing
  • Ideal for mixing blood samples in centrifuge tubes
  • Fixed speed and pitch provide optimum agitation
  • Safe for cold room use
The GyroMini is supplied with a dimpled mat for holding tubes firmly in place.
Informations légales
GyroMini is a trademark of Labnet Intl., Inc.