Principaux documents
Z609641
Bohlender screw caps
GL 18 thread
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.17
Fabricant/nom de marque
Bohlender
Disponibilité
not available in USA and Canada
Raccords
GL 18 thread
Description générale
Made of PPS these screw caps with virgin PTFE membrane gaskets are resistant to high mechanical stresses - even at temperatures upto 250 °C. The gasket consists of PTFE membrane with a multidirectional orientated fibre pattern. Medium will only be exposed to PTFE.
Informations légales
Bohlender is a trademark of Bohlender GmbH