Z549673
High-vacuum valve adapter
male joint: ST/NS 14/20
About This Item
Code UNSPSC :
41121815
Nomenclature NACRES :
NB.35
Produits recommandés
Joint
male joint: ST/NS14/20
Description générale
For high-vacuum applications. Features a high-vacuum PTFE valve. For use with 1/4in. I.D. tubing.