Z506842
Quick Press sample holder
About This Item
Code UNSPSC :
12000000
Nomenclature NACRES :
SB.32
Produits recommandés
Technique(s)
IR spectroscopy: suitable
Description générale
V-shaped sample holder to mount collar containing pellet in 2in. x 3in. slide mount of spectrophotometer.