Key Documents
Z308870
Quickfit® conical flasks
capacity 500 mL, joint: ST/NS 24/29
Synonyme(s) :
FE500/3
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
conical flask
flat bottom
Joint
joint: ST/NS24/29
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
without cap
Conditionnement
case of 5
Fabricant/nom de marque
Quickfit FE500/3
Disponibilité
not available in USA and Canada
Capacité
500 mL
Description générale
With ground-glass socket for general use. Not suitable for vacuum use.
Informations légales
Quickfit is a registered trademark of SciLabware Limited