Principaux documents
Z304808
Quickfit® powder funnels
joint: ST/NS 29/32
Synonyme(s) :
CF5/4
About This Item
Code UNSPSC :
41121809
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS29/32
Fabricant/nom de marque
Quickfit CF 5/4
Disponibilité
not available in USA and Canada
Description générale
For pouring powders or liquids into vessels with ground-glass joints
Informations légales
Quickfit is a registered trademark of SciLabware Limited