Key Documents
Z303356
Quickfit® double surface (Davies) condenser
L 150 mm, joint: ST/NS 19/26
Synonyme(s) :
C5/12/SC
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.43
Joint
joint: ST/NS19/26
Fabricant/nom de marque
Quickfit C 5/12/SC
Disponibilité
not available in USA and Canada
not available in USA and Canada
Longueur
150 mm
Superficie
170 cm2
Description générale
Increased surface area and effectiveness when compared with the corresponding Liebig condenser. Inner cooling surface gives a baffling effect, while outer cooling surface prevents creep and loss of vapour when used with low boiling liquids.
Informations légales
Quickfit is a registered trademark of SciLabware Limited