Principaux documents
Z281352
Laboratory brushes
Centrifuge, L 12 in., brush diam. × L 1 1/2 in. × 4 1/2 in.
About This Item
Code UNSPSC :
47131605
Nomenclature NACRES :
NB.35
Conditionnement
pkg of 4 ea
Longueur
12 in.
brush diam. × L
1 1/2 in. × 4 1/2 in.
Catégories apparentées
Description générale
Brush materials resistant to acids, aromatic and aliphatic hydrocarbons, ketones, ethyl acetate esters, trichloroethylene, and hot water. FDA and USDA approved materials.