Principaux documents
Z170259
Lightnin® 316 stainless steel impeller
3 Blade turbine, Model A-310, diam. 2 1/2 in. , overall
About This Item
Code UNSPSC :
41100000
Nomenclature NACRES :
NB.72
Fabricant/nom de marque
Model A-310
Lightnin 808955
Taille de l'alésage
5/16 in.
Diamètre
2 1/2 in. , overall
Description générale
High efficiency design develops 50% more mixing action and true axial flow.
Informations légales
Lightnin is a registered trademark of Lightnin, a unit of General Signal