Principaux documents
Z137219
Ace round-bottom flask with threaded side-arm
capacity 250 mL
Synonyme(s) :
bottom flask, flask round, round bottom flasks
About This Item
Code UNSPSC :
41121804
Nomenclature NACRES :
NB.43
Matériaux
borosilicate glass
round bottom flask
short
two-neck
Stérilité
non-sterile
Caractéristiques
closure type Standard Taper Joint
Conditionnement
pack of 1
Fabricant/nom de marque
Ace Glass 6936-35
Capacité
250 mL
Description générale
24/40 joint. Side-arm is internally threaded for use with a nylon bushing. Will accommodate thermometers or bleed tubes up to 7 mm diam. Complete with bushing and FETFE® O-ring.
Informations légales
FETFE is a registered trademark of Ace Glass, Inc.