Z136131
Scienceware® heavy-duty pail
capacity 10 L (10.6 qt)
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NB.51
Produits recommandés
Fabricant/nom de marque
Bel-Art
H × D.I.
12 in. × 9 5/8 in.
Capacité
10 L (10.6 qt)
Description générale
PE with large pour spout. Has recessed bottom for better grip while pouring. Graduated inside in L.
Informations légales
Scienceware is a registered trademark of SP Scienceware, a division of SP