Principaux documents
WHA77073500
Whatman® 850-DS 8 channel filter plates
0.2 μm, Nylon membrane, non-sterile, 50 ea
About This Item
Code UNSPSC :
41121800
Nomenclature NACRES :
NC.01
Matériaux:
Nylon membrane
Dimension de pores:
0.2 μm pore size
Type de liaison:
non-treated surface
Nom du produit
Whatman® 850-DS 8-channel filter plate, Nylon membrane, pore size 0.2 μm, pack of 50 ea
Matériaux
Nylon membrane
Conditionnement
pack of 50 ea
Fabricant/nom de marque
Whatman 7707-3500
Dimension de pores
0.2 μm pore size
Type de liaison
non-treated surface
Description générale
Save time and eliminate errors associated with manual sampling. Use Whatman 850-DS 8-channel filter plates in your Agilent 850-DS Dissolution Sampling Station
Application
The filter plates are exclusively designed for use with the optional filter module on the Agilent 850-DS Dissolution Sampling Station to simplify filter replacement between timepoints. Reliable alignment of the liquid path increases productivity in two ways. One, by reducing the risk of jamming, and two, by reducing leaks that may occur with manual sampling or other dissolution sample preparation systems.
Caractéristiques et avantages
  • Automated processing: up to 8 samples simultaneously
  • Readily available: in a wide range of pore sizes and materials
Informations légales
Whatman is a registered trademark of Cytiva