Key Documents
TEM-CF200AUUL
Sigma-Aldrich
Ultrathin Carbon Film Supported Gold Square Mesh
size 200 mesh, box of 50
Synonyme(s) :
CARBON SQUARE MESH - AU 200 MESH UL 50/BX
About This Item
Code UNSPSC :
41111761
Nomenclature NACRES :
MA.24
Conditionnement
box of 50
Taille
200 mesh
Description générale
A 3-4 nm ultrathin film of pure carbon supported standard squre mesh grids. Gold grid, 200 mesh.